missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Erythropoietin/EPO Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB9591-100
This item is not returnable.
View return policy
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| Erythropoietin/EPO | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2056 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Neutralization, ELISA | |
| 148436 | |
| Neutralization 0.15-0.75 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P07321 | |
| EPO | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Erythropoietin/EPO, Ala27-Arg192, Accession # P07321 | |
| 100 μg | |
| Primary | |
| Detects mouse Erythropoietin/EPO in direct ELISAs. In direct ELISAs, approximately 25-50% cross-reactivity with recombinant rat Erythropoietin/EPO, 5-15% cross-reactivity with recombinant human Erythropoietin/EPO, and no cross-reactivity with recombinant mouse Thrombopoietin is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction