missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FGFR2 (IIIc) Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB716-SP
This item is not returnable.
View return policy
Description
FGFR2 Monoclonal specifically detects FGFR2 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| FGFR2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGFR2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse FGF R2 alpha (IIIc) Extracellular domains | |
| 25 μg | |
| Primary | |
| Detects IIIc isoforms of mouse FGF R2 in direct ELISAs and Western blots. Does not react with IIIb isoforms of recombinant mouse (rm) FGF R2. In direct ELISAs and Western blots, 100% cross-reactivity with IIIc isoforms of recombinant human (rh) FGF R2 is observed, and no cross-reactivity with IIIb isoforms of rhFGF R2 or any isoforms of rhFGF R1, rmFGF R3, or rhFGF R4 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Neutralization | |
| 133706 | |
| Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL | |
| BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, FGF R2, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2263 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction