missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Frizzled-7 Antibody, R&D Systems™

Rat Monoclonal Antibody
184.00 € - 1540.00 €
Specifications
| Antigen | Frizzled-7 |
|---|---|
| Clone | 151107 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18728399
![]() |
R&D Systems
MAB198-SP |
25 μg |
184.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18193351
|
R&D Systems
MAB198-050 |
50 μg |
255.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18152361
|
R&D Systems
MAB198-500 |
500 μg |
1540.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Frizzled-7 Monoclonal specifically detects Frizzled-7 in Mouse samples. It is validated for Western Blot.Specifications
| Frizzled-7 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q61090 | |
| 8324 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-7 Gln33-Leu185 (predicted) Accession # Q61090 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects recombinant mouse (rm) Frizzled-7 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Frizzled-1, -2, -3, -4, -6 or -8 is observed. |
| 151107 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| frizzled (Drosophila) homolog 7, frizzled homolog 7 (Drosophila), Frizzled, drosophila, homolog of, 7, Frizzled7, fz-7, FZD7, FzE3frizzled-7, hFz7 | |
| FZD7 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title