missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse G-CSF Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 32 publications
Brand: R&D Systems MAB414-SP
This item is not returnable.
View return policy
Description
G-CSF Monoclonal specifically detects G-CSF in Mouse samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, ELISA Capture (Matched Antibody Pair).
Specifications
| G-CSF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3, CSF3OS, Filgrastim, GCSF, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1440 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, Flow Cytometry, ELISA | |
| 67604 | |
| Western Blot 1 ug/mL, Neutralization 0.01-0.03 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P09920 | |
| CSF3 | |
| E. coli-derived recombinant mouse G-CSF Val31-Ala208 Accession # P09920 | |
| 25 μg | |
| Primary | |
| Detects mouse G-CSF in ELISAs and Western blots. In ELISAs, does not cross-react with recombinant human (rh) G-CSF, rhCNTF, rmIL-6, rmLIF, or rmOSM. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction