missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GFR alpha-3/GDNF R alpha-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF2645-SP
This item is not returnable.
View return policy
Description
GFR alpha-3/GDNF R alpha-3 Polyclonal specifically detects GFR alpha-3/GDNF R alpha-3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| GFR alpha-3/GDNF R alpha-3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GDNF family receptor alpha 3, GDNF family receptor alpha-3, GDNF R alpha-3, GDNF receptor alpha-3, GDNFR-alpha-3, GFR alpha3, GFRa-3, GFRA3, GFR-alpha-3, glial cell line-derived neurotrophic factor receptor alpha-3, GPI-linked receptor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2676 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| AAB70931 | |
| GFRA3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse GFR alpha-3/GDNF R alpha-3 Glu34-Arg379 Accession # AAB70931 | |
| 25 μg | |
| Primary | |
| Detects mouse GFR alpha-3/GDNF R alpha-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human GFR alpha-3 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction