missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GITR/TNFRSF18 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
181.00 € - 423.00 €
Specifications
| Antigen | GITR/TNFRSF18 |
|---|---|
| Clone | 2375B |
| Dilution | Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg;/mL, Immunocytochemistry 8-25 μg;/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18700227
![]() |
R&D Systems
MAB52412-SP |
25 μg |
181.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18724677
|
R&D Systems
MAB52412-100 |
100 μg |
423.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.Specifications
| GITR/TNFRSF18 | |
| Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg;/mL, Immunocytochemistry 8-25 μg;/mL, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| O35714 | |
| 8784.0 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 2375B | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Activation-inducible TNFR family receptor, AITR, AITRTNF receptor superfamily activation-inducible protein, CD357, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, TNFRSF18, tumor necrosis factor receptor superfamily, member 18 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse GITR/TNFRSF18, Accession No. O35714 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse GITR/TNFRSF18 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title