missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Glutaredoxin 1/GLRX1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3119
This item is not returnable.
View return policy
Description
Glutaredoxin 1/GLRX1 Polyclonal specifically detects Glutaredoxin 1/GLRX1 in Mouse samples. It is validated for Western Blot.
Specifications
| Glutaredoxin 1/GLRX1 | |
| Polyclonal | |
| Unconjugated | |
| Q9QUH0 | |
| GLRX | |
| E. coli-derived recombinant mouse Glutaredoxin 1 Met1-Leu107 Accession # Q9QUH0 | |
| 100 μg | |
| Primary | |
| Detects endogenous human and mouse Glutaradoxin in Western blots. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| GLRX, glutaredoxin (thioltransferase), glutaredoxin-1, GRXGRX1, MGC117407, Thioltransferase, Thioltransferase-1, TTase-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2745 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction