missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ICOS Antibody, R&D Systems™

Rabbit Monoclonal Antibody
177.00 € - 438.00 €
Specifications
| Antigen | ICOS |
|---|---|
| Clone | 2317C |
| Dilution | Flow Cytometry 0.25 μg/106 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready |
| Applications | Flow Cytometry, Blocking Assay, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18713087
![]() |
R&D Systems
MAB1682-SP |
25 μg |
177.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18730307
![]() |
R&D Systems
MAB1682-100 |
100 μg |
438.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ICOS Monoclonal specifically detects ICOS in Mouse samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
| ICOS | |
| Flow Cytometry 0.25 μg/106 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| Q9WVS0 | |
| 29851 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 2317C | |
| Flow Cytometry, Blocking Assay, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Activation-inducible lymphocyte immunomediatory molecule, AILIM, AILIMCD278, CD278, CD278 antigen, CRP-1, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
| Chinese Hamster Ovary cell line, CHO-derived mouse ICOS, Met1-Leu142, Accession # Q9WVS0 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse ICOS in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title