Learn More
Mouse IFN-alpha 4 Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB10259-SP
 View more versions of this product
Product Code. 18741257
  • 196.00 € / 25µg
Description
IFN-alpha 4 Monoclonal specifically detects IFN-alpha 4 in Mouse samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-alpha 4
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
IFNA4, IFNalpha 4, IFN-alpha-4, IFN-alpha4a, INFA4, interferon alpha-4, Interferon alpha-4B, Interferon alpha-76, Interferon alpha-M1, interferon, alpha 4, MGC142200
Human embryonic kidney cell HEK293-derived mouse IFN-alpha 4, Leu23-Glu186, Accession # P07351
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG2a
Immunohistochemistry
1020125
Immunohistochemistry 5-25 ug/mL
P07351
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
3441
Mouse
Purified