missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IFN-beta Antibody, R&D Systems™

Rat Monoclonal Antibody
201.00 € - 454.00 €
Specifications
| Antigen | IFN-beta |
|---|---|
| Clone | 944213 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18793227
![]() |
R&D Systems
MAB82341-SP |
25 μg |
201.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742797
![]() |
R&D Systems
MAB82341-100 |
100 μg |
454.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-beta Monoclonal antibody specifically detects IFN-beta in Mouse samples. It is validated for ELISASpecifications
| IFN-beta | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| Human embryonic kidney cell HEK293-derived Mouse IFN-beta , Ile22-Asn182, Accession # P01575 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 944213 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P01575 | |
| 3456 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title