missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgD Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Description
IgD Monoclonal antibody specifically detects IgD in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | IgD |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 11-26C |
| Conjugate | Alexa Fluor 350 |
| Dilution | Flow Cytometry |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Alias | IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta |
| Host Species | Rat |
| Immunogen | Mouse IgD |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?