missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG2a (kappa) Rabbit anti-Mouse, Clone: RM107 (Biotin), Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB14936.50ug
This item is not returnable.
View return policy
Description
Rabbit monoclonal antibody raised against native mouse IgG2a (kappa).
- High specificity
- High reactivity
Specifications
| Mouse IgG2a (kappa) | |
| Monoclonal | |
| Biotin | |
| ELISA (0.005-0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) Immunoprecipitation Western Blotting (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| P01863 | |
| Igh-1a | |
| Native purified mouse IgG. | |
| 50 μg | |
| Primary | |
| Mouse | |
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| RM107 | |
| Rabbit monoclonal antibody raised against native mouse IgG2a (kappa). | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| 1810060O09Rik/IGHG2A/Igh-1/MGC102662/MGC118125/MGC18977 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 380793 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction