missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgM Antibody Pair, Novus Biologicals™
Antibody Pair
463.00 € - 921.00 €
Specifications
| Antigen | Mouse IgM |
|---|---|
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30605613
|
Novus Biologicals
NBP3-48427-5Plates |
5 Plates |
463.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30613118
|
Novus Biologicals
NBP3-48427-15Plates |
15 Plates |
921.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mouse IgM Polyclonal antibody specifically detects Mouse IgM in Mouse samples. It is validated for ELISASpecifications
| Mouse IgM | |
| Polyclonal | |
| RUO | |
| Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH | |
| Storage of components varies. See protocol for specific instructions. |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title