missing translation for 'onlineSavingsMsg'
Learn More

Mouse IL-12/IL-23 p40 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18711997
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18711997 100 μg 100µg
18722187 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18711997 Supplier R&D Systems Supplier No. MAB4992100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-12/IL-23 p40 Monoclonal specifically detects IL-12/IL-23 p40 in Mouse samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12/IL-23 p40
Applications Neutralization
Classification Monoclonal
Clone 2329A
Conjugate Unconjugated
Dilution Neutralization 1-10 ng/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P43432
Gene Alias CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12B, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40
Host Species Rabbit
Immunogen S. frugiperda, Sf 21-derived recombinant mouse IL-12/IL-23 p40 protein, Met1-Ser135, Accession # P43432
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3593
Test Specificity Detects mouse IL-12/IL-23 p40 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.