missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-12/IL-23 p40 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems MAB4991-SP
This item is not returnable.
View return policy
Description
IL-12/IL-23 p40 Monoclonal specifically detects IL-12/IL-23 p40 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-12/IL-23 p40 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Il12b | |
| Recombinant mouse IL-12 p70 heterodimer | |
| 25 μg | |
| Primary | |
| Detects the mouse IL-12/IL-23 p40 subunit. Polymorphisms exist in the mouse p40 sequences [Ymer, S. et al. (2002) Genes and Immunity 3:151.]. | |
| Mouse | |
| Purified |
| Western Blot, Neutralization, ELISA | |
| C17.8 | |
| Western Blot 1 ug/mL, Neutralization 0.04-0.3 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12B, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 16160 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction