Learn More
Mouse IL-22 Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB5821
 View more versions of this product
Product Code. 18718049
  • 512.00 € / 100µg
Description
IL-22 Monoclonal specifically detects IL-22 in Mouse samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-22
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
50616
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
Neutralization
924324
Neutralization 0.5-6 ug/mL
Q9JJY9
IL22
E. coli-derived recombinant mouse IL-22 Leu34-Val179 Accession # Q9JJY9
100 μg
Primary
Detects mouse IL-22 in direct ELISAs.
Mouse
Purified