missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-22 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5821-SP
This item is not returnable.
View return policy
Description
IL-22 Monoclonal specifically detects IL-22 in Mouse samples. It is validated for Neutralization.
Specifications
| IL-22 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 50616 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Neutralization | |
| 924324 | |
| Neutralization 0.5-6 ug/mL | |
| Q9JJY9 | |
| IL22 | |
| E. coli-derived recombinant mouse IL-22 Leu34-Val179 Accession # Q9JJY9 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-22 in direct ELISAs. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction