missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-22 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB582R-SP
This item is not returnable.
View return policy
Description
IL-22 Monoclonal antibody specifically detects IL-22 in Mouse samples. It is validated for Flow Cytometry
Specifications
| IL-22 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
| E. coli-derived recombinant mouse IL-22, Leu34-Val179, Accession # Q9JJY9 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry | |
| 140301R | |
| Intracellular Staining by Flow Cytometry 0.25 ug/mL | |
| Q9JJY9 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 50616 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction