missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LAG-3 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB33281N
This item is not returnable.
View return policy
Description
LAG-3 Monoclonal specifically detects LAG-3 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| LAG-3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Q61790 | |
| LAG3 | |
| NS0 mouse myeloma cell line transfected with mouse LAG-3 Ser23-Leu442 Accession # Q61790 | |
| 100 Tests | |
| 3902 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 631501 | |
| Flow Cytometry 5 uL/10^6 cells | |
| CD223, CD223 antigen, FDC, LAG3, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects mouse LAG-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LAG-3 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction