missing translation for 'onlineSavingsMsg'
Learn More

Mouse Lymphotoxin-alpha/TNF-beta Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

211.00 € - 537.00 €

Specifications

Antigen Lymphotoxin-alpha/TNF-beta
Clone 168717
Dilution Western Blot 1 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18728359
GREENER_CHOICE
View Documents
R&D Systems
MAB749-SP
25 μg
211.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18151311
GREENER_CHOICE
View Documents
R&D Systems
MAB749
500 μg
537.00 €
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lymphotoxin-alpha/TNF-beta Monoclonal specifically detects Lymphotoxin-alpha/TNF-beta in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Lymphotoxin-alpha/TNF-beta
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
LT, LTA, LT-alpha, Lymphotoxin alpha, lymphotoxin alpha (TNF superfamily, member 1), Lymphotoxinalpha, tnfb, TNF-beta, TNFBlymphotoxin-alpha, TNFSF1, TNFSF1B, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1
LTA
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
168717
Western Blot
Unconjugated
Rat
Mouse
P09225
4049
E. coli-derived recombinant mouse Lymphotoxin-alpha/TNF-beta Accession # P09225
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse Lymphotoxin-alpha/TNF-beta in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) TNF-beta , rhTNF-alpha , rmTNF-alpha , rrTNF-alpha , or rcrTNF-alpha is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.