missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Lymphotoxin beta R/TNFRSF3 Antibody, R&D Systems™

Sheep Antibody
218.00 € - 587.00 €
Specifications
| Antigen | Lymphotoxin beta R/TNFRSF3 |
|---|---|
| Dilution | Western Blot 2 μg/mL, Immunohistochemistry 1-15 μg/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18796356
![]() |
R&D Systems
AF10081-SP |
25 μg |
218.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18799526
![]() |
R&D Systems
AF10081-100 |
100 μg |
587.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lymphotoxin beta R/TNFRSF3 specifically detects Lymphotoxin beta R/TNFRSF3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Lymphotoxin beta R/TNFRSF3 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Sheep | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD18, D12S370, ltbetar, LT-BETA-R, LTBR, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, LymphotoxinbR, TNF RIII, TNF Rrp, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
| Mouse myeloma cell line NS0 derived recombinant mouse Lymphotoxin beta Receptor, Ser28-Pro218, Accession # P50284 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Lymphotoxin beta R/TNFRSF3 in Western blots. |
| Western Blot 2 μg/mL, Immunohistochemistry 1-15 μg/mL | |
| Polyclonal | |
| Lyophilized | |
| Mouse | |
| P50284 | |
| 4055 | |
| IgG | |
| Antigen Affinity-purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title