missing translation for 'onlineSavingsMsg'
Learn More

Mouse LYVE-1 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 27 publications

226.00 € - 2155.00 €

Specifications

Antigen LYVE-1
Clone 223322
Concentration LYOPH
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Applications Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18758978
GREENER_CHOICE
View Documents
R&D Systems
MAB2125-SP
25 μg
226.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18726048
GREENER_CHOICE
View Documents
R&D Systems
MAB2125-100
100 μg
546.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18739407
GREENER_CHOICE
View Documents
R&D Systems
MAB2125-500
500 μg
2155.00 €
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Descrizione

Descrizione

LYVE-1 Monoclonal specifically detects LYVE-1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready.
Specifica

Specifica

LYVE-1
LYOPH
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF
Unconjugated
Rat
Mouse
Q8BHC0
10894
BaF/3 mouse pro-B cell line transfected with mouse LYVE-1 Ala24-Thr234 Accession # Q8BHC0
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse LYVE-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD44 or recombinant human LYVE-1 is observed.
223322
Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1XLKD1, XLKD1
LYVE1
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documenti

Documenti

Certificati

For Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato