missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse M-CSF Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB416-SP
This item is not returnable.
View return policy
Description
M-CSF Monoclonal specifically detects M-CSF in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| M-CSF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1435 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Neutralization, ELISA | |
| 131621 | |
| Neutralization 0.3-1.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P07141 | |
| CSF1 | |
| E. coli-derived recombinant mouse M-CSF Lys33-Glu262 (predicted) Accession # P07141 | |
| 25 μg | |
| Primary | |
| Detects mouse M-CSF in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human M-CSF, recombinant mouse (rm) SCF, rmFlt-3 Ligand, rmG-CSF, and rmGM-CSF is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction