missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse NCAM-1/CD56 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
205.00 € - 462.00 €
Specifications
| Antigen | NCAM-1/CD56 |
|---|---|
| Clone | 809220 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18164351
|
R&D Systems
FAB7820G-100UG |
100 μg |
462.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18154531
|
R&D Systems
FAB7820G-025UG |
25 μg |
205.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NCAM-1/CD56 Monoclonal antibody specifically detects NCAM-1/CD56 in Mouse samples. It is validated for Flow CytometrySpecifications
| NCAM-1/CD56 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| CD56, CD56/ NCAM-1, CD56 antigen, MSK39, NCAM1, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
| Mouse myeloma cell line NS0-derived recombinant mouse NCAM-1/CD56, Leu20-Thr711, Accession # P13595 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. |
| 809220 | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Rat | |
| Mouse | |
| P13595 | |
| 4684 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title