missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse NFIL3/E4BP4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
196.00 € - 390.00 €
Specifications
| Antigen | NFIL3 |
|---|---|
| Clone | 1218A |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18729527
![]() |
R&D Systems
MAB8888-SP |
25 μg |
196.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702228
![]() |
R&D Systems
MAB8888 |
100 μg |
390.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NFIL3 Monoclonal specifically detects NFIL3 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| NFIL3 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| E4BP4, E4BP4IL3BP1, IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A | |
| Nfil3 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 1218A | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| O08750 | |
| 4783 | |
| E. coli-derived recombinant mouse NFIL3/E4BP4 Lys140-Arg462 Accession # O08750 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects NFIL3/E4BP4 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title