missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse Nicotinamide N-Methyltransferase/NNMT Recombinant Protein

Product Code. 18200653 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18200653 100 μg 100µg
18217103 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18200653 Supplier Novus Biologicals™ Supplier No. NBP259656100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-264 of Mouse Nicotinamide N-Methyltransferase/NNMT The Recombinant Mouse Nicotinamide N-Methyltransferase/NNMT Protein is derived from E. coli. The Recombinant Mouse Nicotinamide N-Methyltransferase/NNMT Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) PAGE
Formulation In 20mM Tris-HCl (pH8.0) containing 10% glycerol,1mM DTT
Molecular Weight (g/mol) 32.1 kDa
Name Nnmt Recombinant Protein
Purification Method >90% by SDS-PAGE
Quantity 100 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term.
Regulatory Status RUO
Gene Alias FLJ21841, NES, Nestin
Gene Symbol Nnmt
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Muse
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.