missing translation for 'onlineSavingsMsg'
Learn More

Mouse OX40 Ligand/TNFSF4 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18022702
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18022702 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18022702 Supplier R&D Systems Supplier No. AF1236SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

OX40 Ligand/TNFSF4 Polyclonal specifically detects OX40 Ligand/TNFSF4 in Mouse samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OX40 Ligand/TNFSF4
Applications Western Blot, Neutralization, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.2-0.6 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P43488
Gene Alias CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1
Gene Symbols TNFSF4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse OX40 Ligand/TNFSF4 Gln49-Leu198 Accession # P43488
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7292
Test Specificity Detects mouse OX40 Ligand/TNFSF4 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human OX40 Ligand is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.