Learn More
Mouse Plasminogen Kringle 5 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand:  R&D Systems MAB742
 View more versions of this product
Product Code. 18073072
  • 496.00 € / 500µg
Description
Plasminogen Kringle 5 Monoclonal specifically detects Plasminogen Kringle 5 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plasminogen Kringle 5
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Plg
E. coli-derived recombinant mouse Plasminogen Kringle 5 Cys481-Ala563 Accession # AAA50168
500 μg
Primary
Detects mouse Plasminogen Kringle 5 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Plasminogen Kringle 5 is observed.
Mouse
Purified
Western Blot
120304
Western Blot 1 ug/mL
AAA50168
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
5340
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1