missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PlGF-2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB465-500
This item is not returnable.
View return policy
Description
PlGF-2 Monoclonal specifically detects PlGF-2 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| PlGF-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| OORS, PlGF2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5281 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| 62526 | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction | |
| P49764 | |
| Pgf | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse PlGF-2 Ala24-Pro158 Accession # P49764 | |
| 500 μg | |
| Primary | |
| Detects mouse PlGF-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) PlGF or rhVEGF is observed. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction