missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PLZF Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC83951R-100UG
This item is not returnable.
View return policy
Description
PLZF Monoclonal specifically detects PLZF in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| PLZF | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| PLZFzinc finger and BTB domain-containing protein 16, ZBTB16, zinc finger and BTB domain containing 16, Zinc finger protein 145, zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia), Zinc finger protein PLZF, ZNF145, ZNF145Promyelocytic leukemia zinc finger protein | |
| E. coli-derived recombinant mouse PLZF Met1-Gln254 Accession # NP_001028496 | |
| 100 μg | |
| Primary | |
| Detects mouse PLZF in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 816421 | |
| Intracellular Staining by Flow Cytometry | |
| NP_001028496 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7704 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction