missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat CD31/PECAM-1 PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
207.00 € - 355.00 €
Specifications
| Antigen | CD31/PECAM-1 |
|---|---|
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | R-PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18745147
![]() |
R&D Systems
FAB3628P-025 |
25 Tests |
207.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18757387
![]() |
R&D Systems
FAB3628P-100 |
100 Tests |
355.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD31/PECAM-1 Polyclonal specifically detects CD31/PECAM-1 in Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| CD31/PECAM-1 | |
| Flow Cytometry | |
| R-PE | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| PECAM1 | |
| IgG | |
| Affinity Purified | |
| Detects mouse CD31/PECAM-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CD31 and recombinant porcine CD31 is observed. Detects mouse CD31 and rat CD31 in flow cytometry. |
| Flow Cytometry 10 uL/10^6 cells | |
| Polyclonal | |
| Goat | |
| Mouse, Rat | |
| Q08481 | |
| 5175 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD31/PECAM-1 Glu18-Lys590 Accession # Q08481 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title