Learn More
Mouse/Rat RAGE Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 15 publications
Brand:  R&D Systems MAB1179-SP
 View more versions of this product
Product Code. 18705568
  • 242.00 € / 25µg
Description
AGER Monoclonal specifically detects AGER in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAGE
Monoclonal
LYOPH
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
NP_031451
Ager
Mouse myeloma cell line NS0-derived recombinant mouse RAGE Gly23-Ala342 Accession # NP_031451
25 μg
Primary
Detects mouse and rat RAGE in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human RAGE is observed.
Mouse, Rat
Purified
Western Blot, Immunohistochemistry
175410
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
5891
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a