missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat RAGE Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 15 publications
Brand: R&D Systems MAB1179-100
This item is not returnable.
View return policy
Description
RAGE Monoclonal antibody MAB1179100 Specifically detects RAGE in Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
| RAGE | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| NP_031451 | |
| Ager | |
| Mouse myeloma cell line NS0-derived recombinant mouse RAGE Gly23-Ala342 Accession # NP_031451 | |
| 100 μg | |
| Primary | |
| Detects mouse and rat RAGE in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human RAGE is observed. | |
| Mouse, Rat | |
| Supernatant |
| Western Blot, Immunohistochemistry | |
| 175410 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5891 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction