missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Sca-1/Ly6 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1226T-100UG
This item is not returnable.
View return policy
Description
Sca-1/Ly6 Monoclonal specifically detects Sca-1/Ly6 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Sca-1/Ly6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| CAA28351 | |
| LY6A | |
| Mouse myeloma cell line NS0-derived recombinant mouse Sca-1/Ly6 C-terminally truncated Ly-6E allele Leu27-Gly119 Accession # CAA28351 | |
| RUO | |
| 58530 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 177228 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| C6orf23, G6D, Ly6, Ly-6, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25, Sca1 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Sca-1/Ly6 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction