missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SECTM1A Antibody, R&D Systems™

Sheep Antibody
Brand: R&D Systems AF7837-100
This item is not returnable.
View return policy
Description
SECTM1A specifically detects SECTM1A in Mouse samples. It is validated for Immunohistochemistry, Blockade of Receptor-ligand Interaction.
Specifications
| SECTM1A | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| K12, SECTM | |
| Mouse myeloma cell line NS0-derived recombinant mouse SECTM1A Met1-Thr165 Accession # Q921W8 | |
| 100 μg | |
| 6398 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
| Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction 0.2-1 ug/mL | |
| Q921W8 | |
| Sheep | |
| Affinity Purified | |
| Primary | |
| Detects mouse SECTM1A in direct ELISAs. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction