missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Moxifloxacin hydrochloride

Product Code. 15595986 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15595986 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15595986 Supplier Thermo Scientific Chemicals Supplier No. 457960010
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Moxifloxacin hydrochloride, CAS # 186826-86-8, is a broad-spectrum compound with antibacterial activity against both gram-positive and gram-negative bacteria. It also shows inhibitory activity against topoisomerase II.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Moxifloxacin hydrochloride is a fourth-generation synthetic fluoroquinolone compound with antibacterial activity against both gram-positive and gram-negative bacteria

• It also shows an inhibitory activity against topoisomerase II

• Moxifloxacin hydrochloride can bind to and inhibit both bacterial enzymes DNA gyrase (topoisomerase II) and topoisomerase IV, resulting in DNA replication and repair inhibition thus induce cell death in sensitive bacterial species

• Moxifloxacin has 100 times higher affinity for bacterial DNA gyrase compared to mammalian DNA gyrase

Applications

• Moxifloxacin hydrochloride presents a higher anti-gram-positive activity compared to other fluoroquinolone such as ciprofloxacin and ofloxacin

• It has immunosuppressive activity that inhibits the proliferation of all classes of lymphocytes, which can induce apoptosis of antigen-presenting T cells

• It is a cytostatic anthracenedione that can intercalate DNA, increasing the occurence of double-strand breaks by stabilizing the cleavable complex of topoisomerase II and DNA

• This compound shows anticancer activity in some animal models presenting leukemia

• Moxifloxacin is being investigated for the treatment of multidrug-resistant tuberculosis strains

TRUSTED_SUSTAINABILITY

Specifications

Type Moxifloxacin Hydrochloride
CAS 186826-86-8
Melting Point 324.0°C to 325.0°C
Infrared Spectrum Authentic
Water 5% max.
Quantity 1 g
Molecular Formula C21H25ClFN3O4
InChI Key IDIIJJHBXUESQI-DFIJPDEKSA-N
SMILES COC1=C2C(=CC(=C1N3CC4CCCNC4C3)F)C(=O)C(=CN2C5CC5)C(=O)O.Cl
IUPAC Name 7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 437.896
PubChem CID 101526
ChEBI CHEBI:7008
Formula Weight 437.89
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Moxifloxacin hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Moxifloxacin hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Moxifloxacin hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.