missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Spectinomycin, dihydrochloride pentahydrate, PhytoPure™

Product Code. 17610917 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17610917 1 g 1g
17620917 5 g 5g
11470802 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17610917 Supplier MP Biomedicals™ Supplier No. 0219454001
  • ONLINE EXCLUSIVE PRICE
    37.65 €
    35.70 € / 1g
    Save 1.95 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

An aminocyclitol antibiotic, closely related to the aminoglycosides, it inhibits protein synthesis (elongation) by interfering with peptidyl tRNA translocation. It is active against gram-negative bacteria and is used as a selection agent for transformed plant cells that contain the selectable marker gene Spcr.

 

  • Format: Crystalline Powder, Non-sterile
  • Synonyms: U18409AE; Actinospectacin; M141; Trobicin 

Specinomycin is used in cell culture applications, It is active against gram-negative bacteria.It has been shown to have a wide variety of uses, including treating acute gonorrheal urethritis and cervictis, to mark cell layers to monitor cell fate during leaf development, as a selection marker in plant related transformation systems for plant cells containing the marker gene Spcr, to study respiratory tract infections of cattle caused by Pasteurella multocida and Mannheimia haemolytica, and to generate plants deficient for the plastid-encoded RNA polymerase on MS-agar media. Spectinomycin also has use in amplification of low copy number plasmid carrying replicons.

All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 22189-32-8
Content And Storage 4°C
Form Crystalline powder
Product Type Antibiotic
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.