missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FLUCONAZOLE

Product Code. 11406941 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406941 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11406941 Supplier MP Biomedicals™ Supplier No. 0219898650
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective inhibitor of fungal cytochrome P-450 sterol C-14 α-demethylation

A subclass of the synthetic triazole antifungal agents

  • Physical Description: White crystalline powder
  • Molecular Formula: C13H12F2N6O
  • CAS Number: 86386-73-4
  • Synonyms: 2,4-Difluoro-1-bis(1H-1,2,4-triazol-1-ylmethyl) benzyl alcohol; 2,4-Difluoro-α,α1- bis(1H-1,2,4-triazol-1-ylmethyl) benzyl alcohol; α-(2,4-Difluorophenyl)-α-(1H-1,2,4-triazol-1-yl methyl)-1H-1,2,4-triazole-1-ethanol
  • Molecular Weight: 306.27
  • Solubility: Soluble in methanol and chloroform; sparingly soluble in acetone; very slightly to insoluble in water or saline

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.