missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ionomycin

Product Code. 11415691 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415691 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11415691 Supplier MP Biomedicals™ Supplier No. 0215961101
  • ONLINE EXCLUSIVE PRICE
    116.00 €
    98.70 € / 1mg
    Save 17.30 €
    14% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective calcium ionophore

  • Free Acid
  • Molecular Formula: C41H72O9
  • Molecular Weight: 709.0

Specifications

Content And Storage -20°C
Product Type Antibiotic
Quantity 1 mg
compliance-icons
Product Identifier
  • Delta-Aminolevulinic Acid Hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contain: Delta-AminolevulinicAcid Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.