missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Linbro™ Microplate Sealer

Product Code. 11499565 Shop All MP Biomedicals Products
Change view
Click to view available options
No. per Pack:
100
1000
Unit Size:
Pack of 100
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack unitSize
11499565 100 Pack of 100
15702432 1000 Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499565 Supplier MP Biomedicals™ Supplier No. 097640105
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with multi-well plates and tissue culture plates

  • Easy application and removal for all microplates for both short- and long-term storage Provide tight seal to minimize evaporation and condensation
  • Suitable for use between -16 °C and 38 °C
  • Transparent tape and is not pierceable
  • Nonsterile
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Acetate
Sterility Non-sterile
No. per Pack 100
For Use With (Application) Used for multi-well plates and tissue culture plates
Product Type Microplate Sealer
Temperature Range (Metric) -16°C to +38°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.