missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Linbro™ Microplate Sealer

Product Code. 11409575 Shop All MP Biomedicals Products
Change view
Click to view available options
Material:
Mylar
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
11409575 Mylar Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11409575 Supplier MP Biomedicals™ Supplier No. 097640205
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Seal mylar plates wtith these gas-permeable sealers. Mylar sealers are NOT autoclavable.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Mylar
No. per Pack 100
Product Type Microplate Sealer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.