missing translation for 'onlineSavingsMsg'
Learn More

Penicillin G, Sodium Salt, MP Biomedicals™

Product Code. 17600937 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mU
100 mU
25 mU
Unit Size:
1 micro unit
100 micro units
25 micro units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17600937 1 mU 1 micro unit
17620937 25 mU 25 micro units
17630937 100 mU 100 micro units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17600937 Supplier MP Biomedicals Supplier No. 0219453701
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Penicillin G, Sodium Salt Cell culture grade, Powder

  • Penicillin G is a narrow spectrum antibiotic derived from Streptococcus pneumoniae.
  • It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility and in cell culture both alone and combined with streptomycin and other antibiotics.
  • Penicillin G also known as benzylpenicillin.
  • It exert a bacteriocidal as well as a bacteriostatic effect on susceptible bacteria by interfering with the final stages of the synthesis of peptidoglycan, a substance in the bacterial cell wall.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-57-8
MDL Number MFCD00069666
InChI Key FCPVYOBCFFNJFS-LQDWTQKMSA-M
Synonym Benzylpenicillin sodium salt

Specifications

Physical Form Powder
Loss on Drying 0.001
Potency >1500 u/mg
Color White
Quantity 1 mU
Molecular Formula C16H17N2NaO4S
Validated Application Cell Culture
Beilstein 3834217
Solubility Information Soluble in water(H2O):100mg/mL.
Molecular Weight (g/mol) 356.37
Grade Cell Culture
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Penicillin G Sodium Salt, cell culture grade
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Penicillin G SodiumSalt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.