missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Plant Culture Salt Mixtures
Shop All MP Biomedicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 x 10 L
1 x 50 L
Product Type:
Hoagland's Modified Basal Salt Mixture
Murashige and Skoog Basal Salt Mixture
Murashige and Skoog Plant Salt Mixture
missing translation for 'unitSize'
10 L
50 L
Description
- Ready to use
- Liquid formulation, prepared according to the original specification, except where noted
- Available in different pack sizes, ready to use and liquid formulations
Specifications
Specifications
| Form | Powder |
| Product Type | Murashige and Skoog Basal Salt Mixture |
| Quantity | 1 x 10 L |
| Format | Solution |
| pH | 3.5 to 4.5 |
| Solubility | Soluble in water |
| Content And Storage | 4°C |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Reproductive toxicity Category 1A
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H360-May damage fertility or the unborn child.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Potassium Nitrate, Ammonium Nitrate, Calcium Chloride, ZincSulfate (Pentahydrate), Boric Acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction