missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Plant Culture Salt Mixtures

Product Code. 11494895 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 x 10 L
1 x 50 L
Product Type:
Hoagland's Modified Basal Salt Mixture
Murashige and Skoog Basal Salt Mixture
Murashige and Skoog Plant Salt Mixture
Unit Size:
10 L
50 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type Unit Size
11494895 1 x 10 L Hoagland's Modified Basal Salt Mixture 10 L
11405005 1 x 50 L Murashige and Skoog Plant Salt Mixture 50 L
11434925 1 x 10 L Murashige and Skoog Basal Salt Mixture 10 L
11494995 1 x 10 L Murashige and Skoog Plant Salt Mixture 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11494895 Supplier MP Biomedicals™ Supplier No. 092621822
 Restricted Item
Estimated Shipment: 30-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Plant Culture Salt Mixtures are for the molecular testing of plants and plant-derived samples for improving the productivity, quality, and disease resistance of plants.

  • Ready to use
  • Liquid formulation, prepared according to the original specification, except where noted
  • Available in different pack sizes, ready to use and liquid formulations

Specifications

Form Powder
Product Type Hoagland's Modified Basal Salt Mixture
Quantity 1 x 10 L
Format Solution
pH 4.8
Content And Storage 4°C
compliance-icons compliance-icons
Product Identifier
  • Hoagland's Modified Basal Salt Mixture
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Calcium Nitrate Tetrahydrate, PotassiumNitrate, Ferrous Sulfate Heptahydrate, Boric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.