missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Potato Dextrose Broth

Product Code. 11480225 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480225 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11480225 Supplier MP Biomedicals™ Supplier No. 091008617
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suited for the cultivation of a variety of yeasts and molds from dairy products and other foods

Preparation

  • Suspend 24g of the medium in one liter of distilled or deionized water
  • Mix well and heat with frequent agitation
  • Boil for one minute
  • Sterilize at 121°C (15 lb. of pressure) for 15 minutes
  • When the medium is to be used for enumeration of molds and yeasts, add to the medium, sterilized and cooled to 45° to 50°C, approximately 14mL of a sterilized 10% solution of tartaric acid to obtain a pH of 3.5
Physical Appearance
  • Dehydrated medium appearance: Light beige
  • Solubility
    • Soluble in 2.4% distilled or deionized water on boiling

    Potato Dextrose broth can be used in the analysis of dairy products, bottled drinks, frozen food and other types of food. It can also be used in the identification of fungi and yeasts in parallel with their cellular morphology or in methods of mirocultivation in slides.

    Specifications

    Product Type Potato Dextrose Broth
    Quantity 500 g
    pH 4.9 to 5.3
    Content And Storage 15° to 30°C
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.