missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ RPMI 1640 Medium, 1X Liq. with L-Glutamine and Sodium Bicarbonate

Product Code. 11420385 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420385 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11420385 Supplier MP Biomedicals™ Supplier No. 091260354
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Used for the cultivation of normal human and neoplastic leukocytes.

Specifications

Form Liquid
Product Type RPMI 1640
Sterility Sterile
Quantity 500 mL
pH 6.9 to 7.4
Content And Storage 4°C

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.