missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ RPMI 1640 Medium, 1X Liq., w/o L-Glutamine and L-Leucine

Product Code. 11400415 Shop All MP Biomedicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
missing translation for 'unitSize':
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
11400415 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11400415 missing translation for 'mfr' MP Biomedicals™ missing translation for 'supplierNo' 091629149
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for the cultivation of normal human and neoplastic leukocytes. It is now a popular general purpose medium when properly supplemented.

Specifications

Product Type RPMI 1640
Sterility Sterile
Without Additives No Glutamine, No Leucine
Quantity 100 mL
pH 6.9 to 7.5
Content And Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.