missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Sephadex™ G-100

Product Code. 11455222 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g

Product Code. 11455222

Brand: MP Biomedicals™ 0219525510

Fractionation Range (MW)

  • Dry Bead Diameter: 40 to 120μm
  • CAS: 9050-94-6

Specifications

Quantity 10 g
Flow Rate ≥40 cm/hr.
Particle Size 40 to 120 μm
Bed Volume 15 to 20mL/g in Distilled Water
Product Type Sephadex G-100 Media
Format Powder
For Use With (Application) Desalting and group separations of low and high molecular weight molecules, Buffer exchange and the removal of small molecules during the preparation of large biomolecules, such as ampholytes, detergents
Max. Pressure 0.96 bar
Support Type Cross-linked Dextran with Epichlorohydrin
Sample Type Proteins, Enzymes and Peptides
Content And Storage 15° to 30°C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.