Learn More
Novus Biologicals™ Mrc2 Antibody (Quark patent anti-ENDO 180) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28883-1mg
 View more versions of this product
Product Code. 30045423
  • 920.00 € / 1mg
Description
Mrc2 Monoclonal antibody specifically detects Mrc2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mrc2
Monoclonal
Unconjugated
Q9UBG0
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Quark patent anti-ENDO 180
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CD280 antigen, CD280C-type mannose receptor 2, CLEC13EUPAR-associated protein, ENDO180C-type lectin domain family 13 member E, Endocytic receptor 180, KIAA0709FLJ35911, Macrophage mannose receptor 2, mannose receptor, C type 2, UPARAPendocytic receptor (macrophage mannose receptor family), urokinase plasminogen activator receptor-associated protein, Urokinase receptor-associated protein, Urokinase-type plasminogen activator receptor-associated protein
MRC2/ CD280
1 mg
CD Markers, Cell Biology, Immunology
9902
Human
Purified