Learn More
Invitrogen™ MRE11 Recombinant Mouse Monoclonal Antibody (HL1386-MS)
GREENER_CHOICE
Mouse Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556801
Product Code. 30276139
  • 500.00 € / 100µL
Description
MRE11 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
MRE11 (double-strand break repair protein MRE11) is a component of the MRN complex. This complex plays a central role in double-strand break repair, DNA recombination, maintenance of telomere integrity and meiosis. The complex possesses single-stranded endonuclease activity and double-strand-specific 3'-5' exonuclease activity which are provided by MRE11. RAD50 is required to bind DNA ends and hold them in close proximity. Mutations in the gene could result in ataxia-telangiectasia-like disorder 1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MRE11
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P49959, Q61216, Q9JIM0
Mre11, Mre11a
Recombinant fragment of human Mre11.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
HL1386-MS
Unconjugated
Mre11
ATLD; AT-like disease; DNA recombination and repair protein; double-strand break repair protein MRE11; Double-strand break repair protein MRE11A; endo/exonuclease Mre11; HNGS1; Meiotic recombination 11 homolog 1; meiotic recombination 11 homolog A; meiotic recombination 11 homolog A (S. cerevisiae); mmMRE11A; MRE11; MRE11 double strand break repair nuclease A; MRE11 homolog 1; MRE11 homolog A; MRE11 homolog A, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease A; MRE11 meiotic recombination 11 homolog A; MRE11 meiotic recombination 11 homolog A (S. cerevisiae); MRE11 meiotic recombination 11-like protein A; MRE11A; MRE11B; MRX complex nuclease subunit; NGS1; RAD58; XRS4; YM9959.06C; YMR224C
Mouse
Protein A
RUO
17535, 4361, 64046
-20°C, Avoid Freeze/Thaw Cycles